- Volume:833 µl
- Description:Model VDRL 1/60ml LT Syringe
- Cat. No.:60384-104
- Supplier no.:87800
VDRL Antigen Dispensers are used with the PB600-1 Repeating Dispenser (sold separately) to dispense up to 50 times without refilling
Gastight® syringes are ideal for dispensing both liquids and gases. They have a precision-machined PTFE plunger tip which creates a leak-free seal. The fluid path includes SST, borosilicate glass, and PTFE.
With the luer tip termination, needles are removable and fit over a ground glass hub which is tapered in the shape of a male luer. The LT termination will accept most hypodermic needles but was designed specifically for use with Hamilton Kel-F needles. This termination increases the dead volume in the syringe, which may not be appropriate for some applications. Autoclavable to 115°C (239°F) when disassembled.
This qualitative syringe delivers 1/60mL per dispense. A special 90° (bent) syringe needle (sold separately) allows easy syringe reading and filling.