Accessories for METTLER TOLEDO Excellence Benchtop Density Meters, Mettler Toledo
Supplier: METTLER TOLEDO S.A. DE C.V. MX
Certificates
Accessories for METTLER TOLEDO Excellence Benchtop Density Meters, Mettler Toledo
Supplier: METTLER TOLEDO S.A. DE C.V. MX
About this item
Frequently Bought Together
VWR® Mini Centrifuges
Supplier: VWR International
Potassium dihydrogen phosphate ≥99.0% ACS, VWR Chemicals BDH®
Supplier: VWR International
VWR® Traceable® Flashing LED Alert Big-Digit Dual Channel Timer
Supplier: VWR International
PYREX® Graduated Cylinder, To Contain, with PYREX® Stopper, Corning
Supplier: Corning
VWR® Traceable® Four-Channel Alarm Timer with Clock
Supplier: VWR International
VWR® Dot Matrix Printer
Supplier: VWR International
Accessories for Impact Printers
Supplier: Mettler Toledo
KIMTECH SCIENCE® KIMWIPES™ Delicate Task Wipers, KIMBERLY-CLARK PROFESSIONAL®
Supplier: Kimberly-Clark
VWR® Traceable® Three-Line Alarm Timer
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
MICRO-MATE® Luer Lock Syringes, Chemglass
Supplier: Chemglass
2,2,2-Trichloroethanol 99%
Supplier: Thermo Scientific Chemicals
2-Propanol ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Research Chemicals
Buffers, Reference standards, Acidic to Neutral, Ricca Chemical
Supplier: Ricca Chemical
Components for Branson Ultrasonic Cleaners, Branson Ultrasonics
Supplier: Branson Ultrasonics
Buffer Solution, pH 7.00 (±0.01 @ 25 °C), No Color, Specpure®, NIST Traceable, Thermo Scientific
Supplier: Thermo Scientific Chemicals
TRAP SAFE GLASS MOISTURE 1/8 FTG 70ML
Supplier: PerkinElmer