Order Entry
Puerto Rico
ContactUsLinkComponent
GET™ Total RNA for Small Scale, Spin Format RNA Purification, G-Biosciences
GET™ Total RNA for Small Scale, Spin Format RNA Purification, G-Biosciences
Catalog # 95057-558
Supplier:  G-Biosciences
CAS Number:  
undefined
GET™ Total RNA for Small Scale, Spin Format RNA Purification, G-Biosciences
Catalog # 95057-558
Supplier:  G-Biosciences
Supplier Number:  786-132
CAS Number:  

Specifications

  • Method / Format:
    Column (silica-based)
    Organic extraction
  • Prep size:
    Mini
  • Refine target molecule:
    Total RNA
  • Sample type:
    Plant samples
    Bacteria
    Cultured cells
    Human/animal cells and tissues
  • Target molecule:
    RNA
  • Description:
    GET™ Total RNA
  • Size:
    50 Preps
  • Description:
    GET™ Total RNA
  • Cat. No.:
    95057-558
  • Supplier no.:
    786-132

Specifications

About this item

G-Biosciences' GET™ Total RNA kit isolates total RNA from contaminating DNA, proteins, and nucleases using our GET™ RNA Spin Columns.

G-Biosciences' GET™ Total RNA kit contains a simple, less than 60-minute protocol. After homogenization, RNA is bound to the GET™ RNA Spin Columns and washed. The protocol provides an option to remove contaminating DNA with a single DNase treatment. Finally RNA is eluted from the column. The eluted RNA is ready for any procedure including Northern/slot/dot blots, reverse transcription or RNase protective assays.

G-Biosciences' GET™ Total RNA kit rapidly inhibits destructive RNases, using a strong chaotropic buffer for rapid RNA release and high affinity RNA binding spin columns.

G-Biosciences' GET™ Total RNA kit is compatible with numerous species and tissues, including animal and plant tissues, cultured cells, blood and bacteria. The kit is suitable for 50 x 50 mg extractions.

Ordering information: Accessories available.