About this item
For a high PCR workload, the Eppendorf® Mastercycler nexus X2 offers efficiency with two blocks (32-well and 64-well), allowing simultaneous runs of two independent protocols. Accommodates a range of consumables up to 0.5 ml tubes. For increased capacity, connect with up to three additional units.
- Asymmetrically divided block for running different assays in parallel, one 32-well and one 64-well block
- Intuitive graphic programming
- Universal block for strips, 0.1, 0.2 and 0.5 ml PCR tubes and fits for divisible PCR plates
- Small footprint
- Optional gradient for PCR optimization
- E-mail notification
- flexlid® concept allows use of all types of consumables with automatic height adjustment of the lid
- Peltier elements, triple circuit technology
Like every member of the Mastercycler® nexus family, the Mastercycler® nexus X2 can be combined with all other family members in a network of up to three units. In combination with Eppendorf’s PCR tubes, PCR strips or divisible plates, the Mastercycler nexus X2 will give you consistent, publication-ready results - every day!
The flexlid® concept for effortless height adjustment - The flexlid concept enables the nexus X2 PCR thermal cycler to deal with all types of consumables, from low volume all the way up to 0.5 ml PCR tubes. Of course, all common PCR plates fit - and if you ever need higher throughput, you can simply connect one or two Mastercycler® nexus X2 eco units to your Mastercycler® nexus.
Gradient with SteadySlope® technology. The Mastercycler® pro’s gradient option features SteadySlope technology to ensure that heating and cooling ramp rates are identical in both gradient and normal operation. This gives you identical temperature control characteristics in both optimization and routine experiments. Programming the gradient with the Control Panel and the CycleManager pro is easy and intuitive, so that new users can get started quickly and safely.
Specifications
- Catalog No:
- 10119-852
- 10119-850
- 10119-846
- 10119-844
- Model:
- Nexus GX2/GX2e
- Nexus X2/X2e
- Nexus GX2/GX2e
- Nexus X2/X2e
- Block Homogeneity:
- ≤ ±0.3 °C at 20...72 °C
≤ ±0.4 °C at 95 °C - ≤ ±0.3 °C at 20...72 °C
≤ ±0.4 °C at 95 °C - ≤ ±0.3 °C at 20...72 °C
≤ ±0.4 °C at 95 °C - ≤ ±0.3 °C at 20...72 °C
≤ ±0.4 °C at 95 °C
- Block Material:
- Aluminum
- Aluminum
- Aluminum
- Aluminum
- Control Accuracy:
- ±0.2 °C
- ±0.2 °C
- ±0.2 °C
- ±0.2 °C
- Cooling Rate:
- max. 2 °C/s
- max. 2 °C/s
- max. 2 °C/s
- max. 2 °C/s
- Gradient:
- +
- -
- +
- -
- Gradient Temperature Range:
- 30…99 °C
- —
- 30…99 °C
- –
- Heating Rate:
- max. 3 °C/s
- max. 3 °C/s
- max. 3 °C/s
- max. 3 °C/s
- Lid:
- flexlid® technology, with TSP (lid descent)
- flexlid® technology, with TSP (lid descent)
- flexlid® technology, with TSP (lid descent)
- flexlid® technology, with TSP (lid descent)
- Lid Temperature Range:
- 37…110 °C
- 37…110 °C
- 37…110 °C
- 37…110 °C
- Sample capacity:
- 64/32×0.2 ml PCR tubes or up to 24/10×0.5 ml PCR tubes
- 64/32×0.2 ml PCR tubes or up to 24/10×0.5 ml PCR tubes
- 64/32×0.2 ml PCR tubes or up to 24/10×0.5 ml PCR tubes
- 64/32×0.2 ml PCR tubes or up to 24/10×0.5 ml PCR tubes
- Temperature Range:
- 4...99 °C (temp. control range of the block)
- 4...99 °C (temp. control range of the block)
- 4...99 °C (temp. control range of the block)
- 4...99 °C (temp. control range of the block)
- Temperature Uniformity:
- ≤ ±0.4 °C
- ≤ ±0.4 °C
- ≤ ±0.4 °C
- ≤ ±0.4 °C
- W×D×H:
- 25×41.2×33 cm (9.8×16.2×13")
- 25×41.2×33 cm (9.8×16.2×13")
- 25×41.2×33 cm (9.8×16.2×13")
- 25×41.2×33 cm (9.8×16.2×13")