Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Swift™
Swift™ membrane stain
Swift™ membrane stain
Health
Swift™ membrane stain
Catalog # 89167-886
Supplier:
G-Biosciences
CAS Number:
SDS
Expand image
Health
Swift™ membrane stain
Catalog # 89167-886
Supplier:
G-Biosciences
Supplier Number:
786-677
CAS Number:
SDS
Specifications
Size:
20 blots
Cat. No.:
89167-886
Supplier no.:
786-677
Specifications
About this item
Product Details
Documentation
:
:
GHS08
Product Details
Documentation