Order Entry
Puerto Rico
ContactUsLinkComponent
femtoTBST™ Wash Buffer, G-Biosciences
femtoTBST™ Wash Buffer, G-Biosciences
Catalog # 82022-572
Supplier:  G-Biosciences
CAS Number:  
femtoTBST™ Wash Buffer, G-Biosciences
Catalog # 82022-572
Supplier:  G-Biosciences
Supplier Number:  786-161
CAS Number:  

Specifications

  • Description:
    femto TBST™ [10X]
  • Size:
    250 mL
  • Cat. No.:
    82022-572
  • Application:
    Washing Buffer to Replace Standard TBST Buffers
  • Supplier no.:
    786-161

Specifications

About this item

G-Biosciences' femto TBST™ will enhance sensitivity of your immunoassays by minimizing the "washing out" of immuno agents or immuno complexes, a common problem associated with classical TBST buffers

G-Biosciences' femto TBST™ can be used for all your incubation and washing procedures. It is supplied as a 10X solution.

The wash buffers aid in the generation of cleaner backgrounds, resulting in a larger ratio of signal to background.

Ordering information: Accessories available.