Swift™ Film Cleaner, G-Biosciences
Catalog # 89167-888
Supplier: G-Biosciences
Specifications
- Description:Swift™ Film Cleaner
- Environmentally Preferable:
- Cat. No.:89167-888
- Supplier no.:786-678
Specifications
About this item
G-Biosciences' Swift Film Cleaner™ allows researchers to clean film that has been overexposed or has a high background or speckling without having to repeat experiments.
G-Biosciences' Swift Film Cleaner™ is suitable for all experiments that use film to develop the result, including gel shift assays, Western, Southern and Northern blots. The following procedure is suitable for all developed films, new and old, and takes a few minutes to reduce the background. The cleaner is quickly stopped once the correct exposure has been reached.
Ordering information: Accessories available.