Black Phenolic Screw Caps, Rubber Backed/PTFE Faced Liner, Qorpak®
Catalog # 14007-004
Supplier: Avantor
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Cap Size:38-430
- Description:White Rubber-Lined Caps
- Cat. No.:14007-004
- Supplier no.:CAP-00241
Specifications
About this item
Designed for the ultimate in product safety
Autoclavable PTFE provides totally inert inner seal and surface facing the sample or product.