Carbachol 99%
Product Details & Documents
Cholinergic agonist
Purity: 99% CAS No.: 51-83-2 Molecular Formula: H2NCO2CH2CH2N(CH3)3Cl Formula Weight: 182.65 Melting Point: ca 210° dec. Beilstein Reference: 3917459 Merck Reference: 14,1779 UN No.: 2811 MDL No.: MFCD00012011
- CAS Number:51-83-2
- Formula:H₂NC(O)O(CH₂)₂N(CH₃)₃Cl
- MDL Number:MFCD00012011
- Molecular Weight:182.65 g/mol
- Merck Index:14,1779
- Melting Pt:∼ 210 °C (dec.)
- Melting Pt:ca 210 °C (dec.)
- Density:1.389 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:200-127-3
- UN:2811
- ADR:6.1,III
- ADR:"6.1",III
Documents
Certificates
Search by Lot NumberOrganic Building Blocks and Intermediates
Application Notes
VWR GHS Compliance Letter (US)
Additional Product Details
Specifications
Specifications
- Pack type:bottle
- Size:25 g
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



