Order Entry
Puerto Rico
ContactUsLinkComponent
Methoxyacetyl chloride 97% stabilized
Flammable
Corrosive
Irritant
Methoxyacetyl chloride 97% stabilized
Catalog # AAAL03210-14
CAS Number:  
Flammable
Corrosive
Irritant
Methoxyacetyl chloride 97% stabilized
Catalog # AAAL03210-14
Supplier Number:  L03210-14
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    bottle
  • Size:
    25 g
  • Cat. No.:
    AAAL03210-14
  • Supplier no.:
    L03210-14

Specifications

About this item

 :
 :108.52 g/mol
Boiling Pt:112…113 °C
Melting Pt:< –40 °C
Density:1.185
Flash Pt:28 °C(82 °F)
Storage Temperature:Ambient
EINECS:254-169-2
UN:2920
ADR:8,II
 :Stabiised with magnesium oxide (MgO) ∼0.3%
 :GHS02
 :GHS05
 :GHS07
CAS Number:38870-89-2
Formula:C₃H₅ClO₂
MDL Number:MFCD00000728