(±)-2-Methyl-1-butanol 98%
About this item
Purity: 98% CAS No.: 137-32-6 Molecular Formula: C5H12O Formula Weight: 88.15 Flash Point: 43°(109°F) Refractive Index: 1.4110 Melting Point: -70° Boiling Point: 129-131° Beilstein Reference: 1718810 Merck Reference: 14,6030 UN No.: 1105 MDL No.: MFCD00004743
- CAS Number:137-32-6
- Formula:C₅H₁₂O
- MDL Number:MFCD00004743
- Molecular Weight:88.15 g/mol
- Merck Index:14,6030
- Boiling Pt:129…131 °C
- Melting Pt:< –70 °C
- Density:0.815
- Flash Pt:43 °C(109 °F)
- Storage Temperature:Ambient
- EINECS:205-289-9
- UN:1105
- ADR:3,III
3 Options Available Below
Product Details & Documents
Purity: 98% CAS No.: 137-32-6 Molecular Formula: C5H12O Formula Weight: 88.15 Flash Point: 43°(109°F) Refractive Index: 1.4110 Melting Point: -70° Boiling Point: 129-131° Beilstein Reference: 1718810 Merck Reference: 14,6030 UN No.: 1105 MDL No.: MFCD00004743
- CAS Number:137-32-6
- Formula:C₅H₁₂O
- MDL Number:MFCD00004743
- Molecular Weight:88.15 g/mol
- Merck Index:14,6030
- Boiling Pt:129…131 °C
- Melting Pt:< –70 °C
- Density:0.815
- Flash Pt:43 °C(109 °F)
- Storage Temperature:Ambient
- EINECS:205-289-9
- UN:1105
- ADR:3,III
Product Family Options
Product Information
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



