Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Sodium nitrate 98+%

Sodium nitrate 98+%

Oxidizing
Irritant
Sodium nitrate 98+%
Certificates
Sodium nitrate 98+%

3 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Purity: 98+% CAS No.: 7631-99-4 Molecular Formula: NaNO3 Formula Weight: 84.99 Odor: Odorless Melting Point: ca 308° UN No.: 1498 MDL No.: MFCD00011119

  • CAS Number:7631-99-4
  • Formula:NaNO₃
  • MDL Number:MFCD00011119
  • Molecular Weight:84.99 g/mol
  • Merck Index:14,8647
  • Boiling Pt:380 °C (1013 hPa)
  • Melting Pt:∼ 308 °C
  • Density:2.26
  • Storage Temperature:Ambient
  • EINECS:231-554-3
  • UN:1498
  • ADR:5.1,III
  • REACH:01-2119488221-41

Documents

Product Family Options

Product Information

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.