Inert Gas Sampling Syringe Valve, Hamilton Company
Catalog # 65510-025
Supplier: Hamilton
Specifications
- Description:Hamilton Inert Gas Sampling Syringe Valve
- Cat. No.:65510-025
- Supplier no.:86580
Specifications
About this item
Hamilton Company offers the most comprehensive selection of standard and custom syringes and accessories in the industry
The Hamilton MTB Sampling Valve is a lever-actuated on/off valve used for storing and transporting samples in any Gastight® PTFE-Luer Lock (TLL) or Luer Tip (LT) syringe. Convert any TLL or LT syringe into a SampleLock™ syringe using this simple accessory. The syringe attaches to the female luer lock connection leaving the male luer open for connection to any Kel-F® hub. The valves are pressure tested to 100 psig and are autoclavable. The bore I.D. is .039.
Each is manufactured to achieve the highest level of accuracy and precision possible.