- Description:FOCUS™ PhosphoRich™
- Size:5 Preps
- Cat. No.:82022-696
- Supplier no.:786-255
G-Biosciences' FOCUS™ PhosphoRich™ is a ready-to-use kit that enriches phosphorylated proteins and phosphopeptides from complex biological samples
The kit contains spin columns that have a phosphoprotein binding resin with a binding capacity of ~20mg phosphorylated ovalbumin per column. The resin columns supplied with the kit can be reused, if regenerated and stored properly.