ContactUsLinkComponent
FOCUS™ Membrane Proteins, G-Biosciences
FOCUS™ Membrane Proteins, G-Biosciences
Catalog # 82022-676
Supplier:  G-Biosciences
CAS Number:  
undefined
FOCUS™ Membrane Proteins, G-Biosciences
Catalog # 82022-676
Supplier:  G-Biosciences
Supplier Number:  786-249
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    FOCUS™ Membrane Proteins
  • Size:
    50 Preps
  • Cat. No.:
    82022-676
  • Supplier no.:
    786-249
Specifications
About this item
G-Biosciences' FOCUS™ Membrane Proteins is a rapid and highly reproducible method for preparation of membrane or hydrophobic proteins from biological samples for 2D-gel analysis or other applications
The kit is provided with reagents necessary for extraction of membrane proteins and their subsequent preparation for isoelectric focusing or 2D gel resolution for improved spot resolution.
G-Biosciences' FOCUS™ Membrane Proteins kit is designed for >50 preps, where 1 prep is one of the following: 100 mg mammalian tissue, 50μL wet animal cell pellet, 50μL wet yeast pellet, 50μL wet bacteria pellet, or 250 mg plant tissue.
Membrane proteins are extracted with a single step phase partition, with an efficiency greater than 90% with minimal cross-contamination from hydrophilic proteins.