- Environmentally Preferable:
- Cat. No.:89177-044
- Electrical:115 V
- Certification:ETL/ETL-C
- Supplier no.:7701020
With the smallest footprint available, the new CentriVap® micro IR Vacuum Concentrator fits in tight spaces and is ideal for small throughput in molecular biology, proteomics, genomics, genetics, cell biology and drug discovery labs
Everything about the CentriVap® micro IR is small except its many features. Its transparent infrared (IR) glass lid directs heat to the samples to speed evaporation and reduces cross contamination from condensation. The CentriVap® micro IR includes a rotor for twenty-four small samples, a CentriZap® micro IR strobe light to view samples while the rotor is spinning, an built-in diaphragm pump and glass trap everything you need to start concentrating. Simply plug it in and it's ready to use.
Weighing only 20 pounds, this personal-sized concentrator may be easily transported from lab to lab.