ContactUsLinkComponent
Silver D-Stain™ Nucleic Acid & Protein Destain, G-Biosciences
Silver D-Stain™ Nucleic Acid & Protein Destain, G-Biosciences
  82021-352
 :  
undefined
Silver D-Stain™ Nucleic Acid & Protein Destain, G-Biosciences
  82021-352
 :  786-199
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    Silver D-Stain™
  • Size:
    25 mini gels
  • Cat. No.:
    82021-352
  • Supplier no.:
    786-199
G-Biosciences' Silver D-Stain™ can be used for reducing the staining of silver stained gels and revealing over-stained bands
G-Biosciences' Silver D-Stain™ allows you to destain gels completely and restain, or follow other downstream applications such as Western transfer or sequencing.
G-Biosciences' Silver D-Stain™ is suitable for destaining and restaining both silver stained protein gels and nucleic acid gels.
The staining of gels with silver stains often results in over-staining or unevenly stained gels. Silver D-Stain™ eliminates the need to repeat the electrophoresis.