Order Entry
Puerto Rico
ContactUsLinkComponent
Coomassie Brilliant Blue Destaining Solution, G-Biosciences
Coomassie Brilliant Blue Destaining Solution, G-Biosciences
Catalog # 95043-428
Supplier:  G-Biosciences
CAS Number:  
undefined
Coomassie Brilliant Blue Destaining Solution, G-Biosciences
Catalog # 95043-428
Supplier:  G-Biosciences
Supplier Number:  786-499
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Coomassie Brilliant Blue Destaining Solution
  • Size:
    1 L
  • Cat. No.:
    95043-428
  • Supplier no.:
    786-499

Specifications

About this item

After Coomassie Brilliant Blue staining process, the band intensity may be further enhanced by de-staining the stained gel in our CBB De-Staining Solution.

  • Contains Methanol and Acetic acid