ContactUsLinkComponent
InGel™ Array for Protein Digestion, G-Biosciences
InGel™ Array for Protein Digestion, G-Biosciences
Catalog # 82021-380
Supplier:  G-Biosciences
CAS Number:  
undefined
InGel™ Array for Protein Digestion, G-Biosciences
Catalog # 82021-380
Supplier:  G-Biosciences
Supplier Number:  786-241A
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    InGel™ Array
  • Size:
    500 preps
  • Cat. No.:
    82021-380
  • Supplier no.:
    786-241A
Specifications
About this item
G-Biosciences' InGel™ Array is a 96-well format kit suitable for processing larger numbers of protein spots concurrently and is compatible with spot-picking instruments
The protein spots are first excised from the silver stained gel and transferred to a proteomic grade titer plate. Silver stained gel pieces are washed with G-Biosciences' SilverOUT™ to remove inhibitory silver ions. The proteins are then digested within the gel using a Mass Spectrometry Grade Trypsin and a supplied Trypsin Digestion Buffer.
The digested peptides are extracted with G-Biosciences' Pep-Extract™, a high diffusion peptide extraction buffer. The extracted peptides are suitable for mass spectrometry analysis without any subsequent treatments or cleaning procedures.
G-Biosciences' InGel™ Array kit is supplied with reagents sufficient for up to 500 protein spots. The kit includes SilverOUT™ for the removal of silver ions, Trypsin Digestion Buffer for optimal trypsin activity, Pep-Extract™ for high level peptide extraction, and the InGel™ Array titer plates and caps. MSG-Trypsin™ (Mass Spectrometry Grade) trypsin, which increases stability, is also available but sold separately.
Fewer than 96 samples a batch may also be processed without any waste.