BLOK™ BLOTTO, 5% Modified Milk Protein Blocking Solution, G-Biosciences
Catalog # 82022-630
Supplier: G-Biosciences
CAS Number:
Specifications
- Description:BLOK™ BLOTTO 5% non fat milk solution
- Size:2 x 500 mL
- Cat. No.:82022-630
- Application:Non-Fat Dry Milk Solution in TBS
- Supplier no.:786-192
Specifications
About this item
G-Biosciences' BLOK™ BLOTTO is a pre-formulated, convenient, ready-to-use blocking solution
G-Biosciences' BLOK™ BLOTTO contains 5% (w/v) nonfat powdered milk in 25mM Tris and 150mM NaCl, (pH 7.4), with 0.01% Antifoam A, supplied in ready-to-use form.
G-Biosciences' BLOK™ BLOTTO works in about 10 to 15 minutes, and is suitable for Western blots, dot blots, ELISA, immunohistochemistry and other immunochemical applications.
BLOK™ BLOTTO is a 5% novel modified nonfat milk protein solution.
Ordering information: Accessories available.