Optimize PTFE Tubings
Supplier: Optimize
Jump to:
About this item
Good for general/ low pressure applications.
- Various OD and ID options
- Chemically inert to most solvents
- Easy to cut
- Translucent for easy monitoring
13 Options Available Below
Product Details & Documents
Good for general/ low pressure applications.
- Various OD and ID options
- Chemically inert to most solvents
- Easy to cut
- Translucent for easy monitoring
Product Family Options
Product Information
- Ø int.MaterialExt.ØLengthAvailabilityPrice
- 10123-224PTFE tubing, naturalSupplier Number: 10-15-00214
- 21509-040PTFE tubing, naturalSupplier Number: 10-15-00226
- 21509-045PTFE tubing, naturalSupplier Number: 10-15-00238
- 21510-241PTFE tubing, naturalSupplier Number: 10-15-00232
- 21510-588PTFE tubing, naturalSupplier Number: 10-15-00205
- 77100-150PTFE tubing, naturalSupplier Number: 10-15-00220
- 77100-152PTFE tubing, naturalSupplier Number: 10-15-00243
- 21509-039PTFE tubing, naturalSupplier Number: 10-15-00221
- 21509-042PTFE tubing, naturalSupplier Number: 10-15-00227
- 21509-047PTFE tubing, naturalSupplier Number: 10-15-00240
- 21509-051PTFE tubing, naturalSupplier Number: 10-15-00245
- 21510-242PTFE tubing, naturalSupplier Number: 10-15-00233
- 21510-591PTFE tubing, naturalSupplier Number: 10-15-00206