SureBlue Reserve™ provides maximum sensitivity for detection of peroxidase labeled conjugates in a microwell format and provides 50% more sensitivity than standard SureBlue™ TMB substrate
- Ready-to-use formulation
The substrate develops a deep blue color that is measurable at 650 nm and turns yellow when stopped by acidification and read at 450 nm. Recommended for ELISA. Single component, ready to use format of 3,3', 5, 5'-Tetramethylbenzidine. No reagent preparation required, very convenient. Stable for 2 years from data of manufacture when stored at 4oC.