


Imidazole
Catalog # 101097-040
Supplier: USP
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Bottle
- Size:200 mg
- Cat. No.:101097-040
- Supplier no.:1336500
Specifications
About this item
:
Merck Index:13,04935
Boiling Pt:255 °C (1013 hPa)
Boiling Pt:255 °C (1013 hPa)
Melting Pt:89…90 °C
Melting Pt:89 - 90 °C
Density:1.18 g/cm³ (20 °C)
Flash Pt:135 °C
Flash Pt:135 °C
Storage Temperature:Ambient
EINECS:206-019-2
UN:2923
ADR:8,III
:68.08 g/mol
:GHS05
:GHS08
:GHS07
CAS Number:288-32-4
Formula:C₃H₄N₂
MDL Number:MFCD00005183