Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Gemfibrozil

Bottle, 200 mg
Catalog # 101095-072
Marketsource
Supplier:   USP
 :  1288500
Gemfibrozil
Catalog # 101095-072
Supplier:  USP
Marketsource
CAS Number:  25812-30-0
Gemfibrozil
Gemfibrozil

Bottle, 200 mg

CAS Number:  25812-30-0
Price
$398.46
Log in for availability
Each (200mg)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  5-(2,5-Dimethylphenoxy)-2,2-dimethylpentanoic acid, 2,2-Dimethyl-5-(2,5-dimethylphenoxy)pentanoic acid, CI-719, 2,2-Dimethyl-5-(2,5-dimethylphenoxy)valeric acid
  • CAS Number:25812-30-0
  • Formula:C₁₅H₂₂O₃
  • MDL Number:MFCD00079335
  • Molecular Weight:250.34
  • Merck Index:12,04394
  • Boiling Pt:395 °C (1013 hPa)
  • Melting Pt:59…61 °C
  • EINECS:247-280-2
Specifications

Specifications

  • Pack type:
    Bottle
  • Size:
    200 mg

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.