Order Entry
Puerto Rico
ContactUsLinkComponent
Dextran, M.W. ~ 70,000 system suitability standard
Dextran, M.W. ~ 70,000 system suitability standard
Catalog # 101094-776
Supplier:  USP
CAS Number:  
undefined
Dextran, M.W. ~ 70,000 system suitability standard
Catalog # 101094-776
Supplier:  USP
Supplier Number:  1179763
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Bottle
  • Size:
    200 mg
  • Cat. No.:
    101094-776
  • Supplier no.:
    1179763

Specifications

About this item

 :
EINECS:232-677-5
CAS Number:9004-54-0
Formula:(C₆H₁₀O₅)n
MDL Number:MFCD00130935