Jump to:
Product Details & Documents
Synonyms: 3α,7α,12α-Trihydroxy-5β-cholan-24-oic acid, 3α,7α,12α-Trihydroxycholan-24-oic acid, (R)-4-((3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-Trihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanoic acid, 3α,7α,12α-Trihydroxycholanic acid, 3α,7α,12α-Trihydroxy-5β-cholanic acid
- CAS Number:81-25-4
- Formula:C₂₄H₄₀O₅
- MDL Number:MFCD00003672
- Molecular Weight:408.58
- Merck Index:13,02223
- Boiling Pt:545 °C (1013 hPa)
- Melting Pt:199 °C
- Storage Temperature:Ambient
- EINECS:201-337-8
Documents
10259592.pdf
SDS
Specifications
Specifications
- Pack type:Bottle
- Size:2 g
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



