Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Calcium hydroxide

Calcium hydroxide

Bottle, 1 g
Catalog # 101094-468
Corrosive
Irritant
Marketsource
Supplier:   USP
Supplier #:  1086855
Calcium hydroxide
Catalog # 101094-468
Supplier:  USP
Marketsource
CAS Number:  1305-62-0
Calcium hydroxide
Calcium hydroxide

Bottle, 1 g

CAS Number:  1305-62-0
Price
Inquire For Price
Log in for availability
Each (1g/mol)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  Slaked lime, Calcium dihydroxide
  • CAS Number:1305-62-0
  • Formula:Ca(OH)₂
  • MDL Number:MFCD00010901
  • Molecular Weight:74.09 g/mol
  • Merck Index:13,01675
  • Melting Pt:550 °C
  • Density:2.24 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-137-3
  • UN:3262
  • ADR:8,III

Documents

Specifications

Specifications

  • Pack type:
    Bottle
  • Size:
    1 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.