Order Entry
Puerto Rico
ContactUsLinkComponent
2,6-Di-tert-butyl-p-cresol
2,6-Di-tert-butyl-p-cresol
Catalog # 101094-446
Supplier:  USP
CAS Number:  
undefined
2,6-Di-tert-butyl-p-cresol
Catalog # 101094-446
Supplier:  USP
Supplier Number:  1082708
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Bottle
  • Size:
    500 mg
  • Cat. No.:
    101094-446
  • Supplier no.:
    1082708

Specifications

About this item

 :
 :220.35 g/mol
Merck Index:13,01547
Boiling Pt:265 °C (1013 hPa)
Melting Pt:69…70 °C
Density:1.05 g/cm³ (20 °C)
Flash Pt:127 °C
Storage Temperature:Ambient
EINECS:204-881-4
CAS Number:128-37-0
Formula:C₁₅H₂₄O
MDL Number:MFCD00011644