Order Entry
Puerto Rico
ContactUsLinkComponent
4-Aminobenzoic acid
4-Aminobenzoic acid
Catalog # 101094-262
Supplier:  USP
CAS Number:  
undefined
4-Aminobenzoic acid
Catalog # 101094-262
Supplier:  USP
Supplier Number:  1019803
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Bottle
  • Size:
    200 mg
  • Cat. No.:
    101094-262
  • Supplier no.:
    1019803

Specifications

About this item

 :
 :137.14 g/mol
Merck Index:13,00422
Boiling Pt:340 °C (1013 hPa)
Melting Pt:188 °C
Density:1.45 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:205-753-0
CAS Number:150-13-0
Formula:C₇H₇NO₂
MDL Number:MFCD00007894