Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Aluminium hydroxide

Aluminium hydroxide

Bottle, 200 mg
Catalog # 101094-246
Irritant
Marketsource
Supplier:   USP
Supplier #:  1017502
Aluminium hydroxide
Catalog # 101094-246
Supplier:  USP
Marketsource
CAS Number:  21645-51-2
Aluminium hydroxide
Aluminium hydroxide

Bottle, 200 mg

CAS Number:  21645-51-2
Price
$415.38
Log in for availability
Each (200mg)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  Hydrargillite, Aluminum hydroxide
  • CAS Number:21645-51-2
  • Formula:Al(OH)₃
  • MDL Number:MFCD00003420
  • Molecular Weight:78 g/mol
  • Merck Index:12,00355
  • Melting Pt:300 °C
  • Density:2.42 g/cm³ (20 °C)
  • EINECS:244-492-7

Documents

Specifications

Specifications

  • Pack type:
    Bottle
  • Size:
    200 mg

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.