Order Entry
Puerto Rico
ContactUsLinkComponent
Ethanol 100% Dehydrated, Reference Standard, USP
Ethanol 100% Dehydrated, Reference Standard, USP
Catalog # 101094-232
Supplier:  USP
CAS Number:  
undefined
Ethanol 100% Dehydrated, Reference Standard, USP
Catalog # 101094-232
Supplier:  USP
Supplier Number:  1012772
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    5 ampoules
  • CAS No.:
    64-17-5
  • Description:
    Ethanol 100% Dehydrated USP Reference Standard
  • Size:
    1200 µl
  • Formula:
    CH₃CH₂OH
  • Description:
    Ethanol 100% Dehydrated
  • Cat. No.:
    101094-232
  • Supplier no.:
    1012772

Specifications

About this item

  • Total alcohol: ~100%
  • Ethyl alcohol: ~100%
  • Denaturant: none