Order Entry
Puerto Rico
ContactUsLinkComponent
Zinc selenide, crystals (parallelogram) 60 ° for spectrophotometry
Toxic
Health
Environment
Zinc selenide, crystals (parallelogram) 60 ° for spectrophotometry
Catalog # 100213-218
Minority or Woman-Owned Business Enterprise
CAS Number:  
undefined
Toxic
Health
Environment
Zinc selenide, crystals (parallelogram) 60 ° for spectrophotometry
Catalog # 100213-218
Supplier Number:  0003J-3073
Minority or Woman-Owned Business Enterprise
CAS Number:  

Specifications

  • Size:
    52.5×20×3 mm
  • Cat. No.:
    100213-218
  • Supplier no.:
    0003J-3073

Specifications

About this item

 :
 :144.35 g/mol
Merck Index:12,10290
Melting Pt:1793 °C
Density:5.4 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-259-7
UN:3283
ADR:6.1,III
 :GHS06
 :GHS08
 :GHS09
CAS Number:1315-09-9
Formula:SeZn
MDL Number:MFCD00011296