Order Entry
United States
ContactUsLinkComponent
Potassium chloride, crystal cubes 1 cm for spectrophotometry
Potassium chloride, crystal cubes 1 cm for spectrophotometry
Catalog # 100213-938
Minority or Woman-Owned Business Enterprise
CAS Number:  
undefined
Potassium chloride, crystal cubes 1 cm for spectrophotometry
Catalog # 100213-938
Supplier Number:  0002-7391
Minority or Woman-Owned Business Enterprise
CAS Number:  

Specifications

  • Size:
    227 g
  • Environmentally Preferable:
  • Cat. No.:
    100213-938
  • Supplier no.:
    0002-7391

Specifications

About this item

Synonyms: Potassiumchloride
 :
 :74.55 g/mol
Merck Index:14,07621
Boiling Pt:1420 °C (1013 hPa)
Boiling Pt:1420 °C (1013 hPa)
Melting Pt:773 °C
Density:1.98 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-211-8
CAS Number:7447-40-7
Formula:KCl
MDL Number:MFCD00011360