


Zinc selenide, crystal disk for spectrophotometry, polished
Supplier: International Crystal Labs
Certificates
About this item
CAS Number:1315-09-9
:
Formula:SeZn
MDL Number:MFCD00011296
:144.35 g/mol
Merck Index:12,10290
Melting Pt:1793 °C
Density:5.4 g/cm³ (20 °C)
EINECS:215-259-7
UN:3283
ADR:6.1,III
:GHS06
:GHS08
:GHS09
Storage Temperature:Ambient
Frequently Bought Together