Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Sodium perborate monohydrate for synthesis

Sodium perborate monohydrate for synthesis

Oxidizing
Corrosive
Health
Irritant
Sodium perborate monohydrate for synthesis
Certificates
Sodium perborate monohydrate for synthesis

4 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Sodium Perborate, Monohydrate is also known PBS-1, is used as a source of active oxygen in cleaning products and detergents. It has also been used as a tooth bleaching formula.

Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

  • CAS Number:10332-33-9
  • Formula:BH₂NaO₄·1H₂O
  • MDL Number:MFCD00149230
  • Molecular Weight:117.83 g/mol
  • Storage Temperature:Ambient
  • EINECS:239-172-9
  • UN:3377
  • ADR:5.1,III

Documents

Certificates
Search by Lot Number
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Poly bottle
    500 g
    Log in for availability
    Log in for availability
    Each (500g)
    Each (500g) - $212.75
    $212.75
    Each (500g)
       $212.75
    Minimum order quantity is 1

    Supplier Number: S1589-500GM
  • Poly bottle
    2.5 kg
    Log in for availability
    Log in for availability
    Each (2,500g)
    Each (2,500g) - $559.41
    $559.41
    Each (2,500g)
       $559.41
    Minimum order quantity is 1

    Supplier Number: S1589-2.5KG
  • Poly pail
    12 kg
    Log in for availability
    Log in for availability
    Each (12kg)
    Each (12kg) - $1,253.97
    $1,253.97
    Each (12kg)
       $1,253.97

    Supplier Number: S1589-12KG
  • Fiber drum
    50 kg
    Log in for availability
    Log in for availability
    Each (50kg)
    Each (50kg) - $2,221.98
    $2,221.98
    Each (50kg)
       $2,221.98

    Supplier Number: S1589-50KGBL

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.