Order Entry
Puerto Rico
ContactUsLinkComponent
Sodium hypochlorite 12% available chlorine in aqueous solution, Reagent Grade
Corrosive
Environment
Sodium hypochlorite 12% available chlorine in aqueous solution, Reagent Grade
Catalog # 700001-340
CAS Number:  
Corrosive
Environment
Sodium hypochlorite 12% available chlorine in aqueous solution, Reagent Grade
Catalog # 700001-340
Supplier Number:  S1316-200LTB
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Drum
  • Size:
    200 L
  • Cat. No.:
    700001-340
  • Supplier no.:
    S1316-200LTB

Specifications

About this item

 :
 :74.44 g/mol
Boiling Pt:111 °C (1013 hPa)
Melting Pt:–30…–20 °C
Density:1.206
Storage Temperature:Ambient
EINECS:231-668-3
UN:1791
ADR:8,III
 :GHS05
 :GHS09
CAS Number:7681-52-9
Formula:ClNaO
MDL Number:MFCD00011120