
Acetonylacetone ≥97%
Supplier: Spectrum Chemical Mfg. Corp.
Certificates
About this item
CAS Number:110-13-4
:
Formula:C₆H₁₀O₂
MDL Number:MFCD00008792
:114.14 g/mol
Merck Index:13,00072
Boiling Pt:191 °C (1013 hPa)
Melting Pt:-5.4 °C
Density:0.971 g/cm³ (20 °C)
Flash Pt:65 °C
Storage Temperature:Ambient
EINECS:203-738-3
UN:2810
ADR:6.1,III
:GHS06
Specification Test Results
- Assay:97% Min.