Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
(±)-1,2-Hexanediol ≥98% for synthesis

(±)-1,2-Hexanediol ≥98% for synthesis

(±)-1,2-Hexanediol ≥98% for synthesis
Certificates

3 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

DL-1,2-Hexanediol, also known as 1,6-Hexanediol or Hexamethylene glycol, can be used for polyurethane production and industrial polyester.

Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Synonyms:  Hexane-1,2-diol, DL-1,2-Hexanediol, (±)-1,2-Dihydroxy hexane
  • CAS Number:6920-22-5
  • Formula:C₆H₁₄O₂
  • MDL Number:MFCD00010737
  • Molecular Weight:118.18 g/mol
  • Boiling Pt:223 °C (1013 hPa)
  • Melting Pt:45 °C
  • Density:0.951
  • Storage Temperature:Ambient
  • EINECS:230-029-6

Documents

Certificates
Search by Lot Number
Specifications for Product Family

Specification Test Results

  • Assay:
    98% Min.
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Glass, amber
    100 g
    Log in for availability
    Log in for availability
    Each (100g)
    Each (100g) - $183.01
    $183.01
    Each (100g)
       $183.01

    Supplier Number: H2008-100GM
  • Poly drum
    20 kg
    Log in for availability
    Log in for availability
    Each (20kg)
    Each (20kg) - $6,443.59
    $6,443.59
    Each (20kg)
       $6,443.59

    Supplier Number: H2008-20KGBL
  • Drum
    200 kg
    Log in for availability
    Log in for availability
    Each (200kg)
    Each (200kg)

    Supplier Number: H2008-200KGB

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.