Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Hydrogen peroxide solution 30% in aqueous solution ACS

Hydrogen peroxide solution 30% in aqueous solution ACS

Corrosive
Irritant
Hydrogen peroxide solution 30% in aqueous solution ACS
Certificates
Hydrogen peroxide solution 30% in aqueous solution ACS

4 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Hydrogen Peroxide Solution, 30 Percent, Reagent, ACS, is the simplest peroxide, a strong oxidizer that appears as a clear and slightly viscous liquid. Its oxidizing properties are so strong that it is considered a highly reactive oxygen species. Due to it being a weak acid it can form hydroperoxide or peroxide salts. One of its biggest uses is in the treatment of drinking water, removing odors, organic materials and the amount of Hydrogen sulfides and Iron. As an ACS grade Reagent, Spectrum Chemical manufactured Hydrogen Peroxide is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness

  • CAS Number:7722-84-1
  • Formula:H₂O₂
  • MDL Number:MFCD00011333
  • Molecular Weight:34.01
  • Boiling Pt:∼107 °C (1013 hPa)
  • Melting Pt:< 0 °C
  • Density:1.11 g/cm³ (20 °C)
  • Storage Temperature:Refrigerator
  • EINECS:231-765-0
  • UN:2014
  • ADR:5.18,II
  •  :Density: 1.11 g/cm³ (20 °C)

Documents

Certificates
Search by Lot Number
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Poly bottle
    100 ml
    Log in for availability
    Log in for availability
    Each (100ml)
    Each (100ml) - $150.34
    $150.34
    Each (100ml)
       $150.34

    Supplier Number: H1065-100ML
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Poly bottle
    500 ml
    Log in for availability
    Log in for availability
    Each (500ml)
    Each (500ml) - $400.94
    $400.94
    Each (500ml)
       $400.94

    Supplier Number: H1065-500MLP
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Poly bottle
    4 L
    Log in for availability
    Log in for availability
    Each (4l)
    Each (4l) - $970.23
    $970.23
    Each (4l)
       $970.23

    Supplier Number: H1065-4LTPL
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Drum
    200 L
    Log in for availability
    Log in for availability
    Each (200l)
    Each (200l) - $4,231.99
    $4,231.99
    Each (200l)
       $4,231.99

    Supplier Number: H1065-200LTB
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.