Fumaric acid ≥99.5% NF suitable for use as excipient
About this item
Fumaric Acid, NF is used an effervescent agent in tablet formulations. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
- CAS Number:110-17-8
- Formula:C₄H₄O₄
- MDL Number:MFCD00002700
- Molecular Weight:116.07 g/mol
- Merck Index:13,04308
- Boiling Pt:356 °C (1013 hPa)
- Melting Pt:287 °C
- Density:1.625 g/cm³ (20 °C)
- Flash Pt:273 °C
- Storage Temperature:Ambient
- EINECS:203-743-0
3 Options Available Below
Product Details & Documents
Fumaric Acid, NF is used an effervescent agent in tablet formulations. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
- CAS Number:110-17-8
- Formula:C₄H₄O₄
- MDL Number:MFCD00002700
- Molecular Weight:116.07 g/mol
- Merck Index:13,04308
- Boiling Pt:356 °C (1013 hPa)
- Melting Pt:287 °C
- Density:1.625 g/cm³ (20 °C)
- Flash Pt:273 °C
- Storage Temperature:Ambient
- EINECS:203-743-0
Documents
Specifications for Product Family
Specification Test Results
- Assay:99.5% Min.
Product Family Options
Product Information
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



