
Ammonium chloride ≥99.5%, granular USP suitable for use as excipient
Catalog # 95031-826
Supplier: Spectrum Chemical Mfg. Corp.
Small Business Enterprise
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Poly bottle
- Size:500 g
- Cat. No.:95031-826
- Supplier no.:AM175-500GM
Specifications
Specification Test Results
- Assay:99.5% Min.
Specification Test Results
About this item
:
:53.49 g/mol
Merck Index:13,00510
Melting Pt:335 °C
Density:1.5256 g/cm³ (20 °C)
EINECS:235-186-4
:GHS07
Storage Temperature:Ambient
CAS Number:12125-02-9
Formula:NH₄Cl
MDL Number:MFCD00011420