Ammonium thiocyanate, crystals ACS for synthesis
About this item
Ammonium Thiocyanate, Crystal, Reagent, ACS is an inorganic laboratory reagent commonly used by the soft drinks industry to determine the iron content in soft drinks by colorimetry.
As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.
- CAS Number:1762-95-4
- Formula:NH₄SCN
- MDL Number:MFCD00011428
- Molecular Weight:76.12 g/mol
- Merck Index:13,00564
- Melting Pt:150 °C
- Density:1.305 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:217-175-6
6 Options Available Below
Product Details & Documents
Ammonium Thiocyanate, Crystal, Reagent, ACS is an inorganic laboratory reagent commonly used by the soft drinks industry to determine the iron content in soft drinks by colorimetry.
As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.
- CAS Number:1762-95-4
- Formula:NH₄SCN
- MDL Number:MFCD00011428
- Molecular Weight:76.12 g/mol
- Merck Index:13,00564
- Melting Pt:150 °C
- Density:1.305 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:217-175-6
Specifications for Product Family
Specification Test Results
- Assay:97.5% Min.
Product Family Options
Product Information
- Pack typeSizeAvailabilityPrice
- Supplier Number: A1270-25GM
- Supplier Number: A1270-125GM
- Supplier Number: A1270-500GM
- Supplier Number: A1270-2.5KG
- Supplier Number: A1270-12KG
- Supplier Number: A1270-50KGBL
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



