Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Vanadium(III) oxide ≥95%

Vanadium(III) oxide ≥95%

Toxic
Health
Irritant
Marketsource
Supplier:   Strem Chemicals Inc
Vanadium(III) oxide ≥95%
Marketsource
Certificates
Vanadium(III) oxide ≥95%

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
  • CAS Number:1314-34-7
  • Formula:O₃V₂
  • MDL Number:MFCD00011455
  • Molecular Weight:149.88
  • Merck Index:12,10059
  • Melting Pt:1970 °C
  • Density:4.87
  • EINECS:215-230-9
  • UN:3285
  • ADR:6.1,II
  •  :Density: 4.87 g/cm³ (20 °C)
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Glass bottle
    50 g
    Log in for availability
    Log in for availability
    Each (50g)
    Each (50g) - $45.29
    $45.29
    Each (50g)
       $45.29

    Supplier Number: 93-2312-50G
    Specifications:
    Marketsource
    Marketsource
  • Glass bottle
    250 g
    Log in for availability
    Log in for availability
    Each (250g)
    Each (250g) - $179.75
    $179.75
    Each (250g)
       $179.75

    Supplier Number: 93-2312-250G
    Specifications:
    Marketsource
    Marketsource

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.