Titanium ≥99.97%, wire
Marketsource
Supplier: Strem Chemicals Inc
Jump to:
About this item
- CAS Number:7440-32-6
- Formula:Ti
- MDL Number:MFCD00011264
- Molecular Weight:47.88 g/mol
- Merck Index:12,09610
- Boiling Pt:3262 °C (1013 hPa)
- Melting Pt:1660 °C
- Density:4.5
- EINECS:231-142-3
2 Options Available Below
Product Details & Documents
- CAS Number:7440-32-6
- Formula:Ti
- MDL Number:MFCD00011264
- Molecular Weight:47.88 g/mol
- Merck Index:12,09610
- Boiling Pt:3262 °C (1013 hPa)
- Melting Pt:1660 °C
- Density:4.5
- EINECS:231-142-3
Product Family Options
Product Information
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



