Specifications
- Description:Column-PROTEIN-Concentrate™
- Sample Size:For 4 mg protein
- Cat. No.:82021-284
- Application:Chromatographic Capture and Elution that Protects Biological Activity
- Supplier no.:786-126
Specifications
About this item
The Column-PROTEIN-Concentrate™ kit has been specifically developed for concentration of those proteins that cannot be concentrated either by precipitation or other techniques
The immobilized protein is spin-eluted in a small volume of specifically formulated elution buffer, giving several fold effective concentration. The method is gentle and protects protein from denaturation during the concentration process. The Column-PROTEIN-Concentrate™ kit maintains and protects the biological activity of proteins and allows for ~100% recovery.
This kit is suitable for concentration of a total of 4mg protein in either single or multiple procedures. For concentration of larger than 5mg protein, a custom made kit may be ordered. The kit is supplied with a proprietary spin column; the column may be regenerated and reused once.
The kit is based on a proprietary Protein Binding Resin that binds and immobilizes any protein in a low salt buffer between pH 2-12. The binding capacity is ~0.5mg protein/ml Protein Binding Resin.