Specifications
- Description:OrgoSol-PROTEIN-Concentrate™
- Sample Volume:For 5 mL protein
- Cat. No.:82021-280
- Application:Organic Solvent Buffer Precipitation Suitable to Protect Biological Activity
- Supplier no.:786-125
Specifications
About this item
The OrgoSol-PROTEIN-Concentration™ kit precipitates protein with a proprietary solvent buffer, OrgoSol™
The kit has been extensively tested for the concentration of a wide selection of enzymatic proteins without the loss of their biological activity and for ~100% protein recovery. The kit is designed to precipitate up to 5ml protein solution.
The method involves mixing a protein solution with the OrgoSol™ Buffer followed by incubation, which results in quantitative precipitation of the protein. The precipitated protein is suspended in a smaller volume of an appropriate buffer and the concentrated protein solution is ready for use.
The OrgoSol™ buffer has been specifically developed for efficient precipitation of protein solutions with minimal disruption to the protein structure and therefore maintains the biological activity of most proteins.