ContactUsLinkComponent
ProteSEEKER™, Protease Estimation Assay, G-Biosciences
  78000-084
 :  
undefined
ProteSEEKER™, Protease Estimation Assay, G-Biosciences
  78000-084
 :  786-325
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    ProteSEEKER™
  • Size:
    50 assays
  • Cat. No.:
    78000-084
  • Supplier no.:
    786-325
ProteSEEKER™ identifies specific types of proteases with a panel of twelve protease inhibitors and a sensitive colorimetric protease screening assay
ProteSEEKER™ protease screening assay consists of a ready-to-use dye-labeled protein, which is digested by proteases to release dye-labeled peptides. The absorbance of which is measured for determination of protease activity. The inhibitors are supplied at a 100X concentration and the 1X concentration provides >90% inhibition in most biological samples.
Accessories: Resorufin-Casein Protease Substrate (#786-321): A colorimetric substrate that when treated with proteases releases resorufin that has an absorbance of 570nm. Supplied lyophilized. FITC-Casein Protease Substrate (#786-322): A fluorescent (fluorescein isothiocyanate (FITC)) substrate that when treated with proteases releases FITC that has an excitation at 485nm and emission at 535nm. Supplied lyophilized.
ProteSEEKER™ allows researchers to screen their protein samples and establish which specific class of proteases are present and therefore design a highly specific protease inhibitor cocktail using the minimal number of protease inhibitors. Alternatively, ProteSEEKER™ can be used to test existing protease inhibitor cocktails and identify their inadequacies and therefore supplement in additional protease inhibitors.
 : Accessories not included.