Order Entry
Puerto Rico
ContactUsLinkComponent
Ethanol denatured ≥100% DLI SDA 3A (200 Proof) (denatured with 5% isopropanol and 5% methanol)
Flammable
Ethanol denatured ≥100% DLI SDA 3A (200 Proof) (denatured with 5% isopropanol and 5% methanol)
Catalog # 71002-398
Supplier:  Decon Labs
CAS Number:  
Flammable
Ethanol denatured ≥100% DLI SDA 3A (200 Proof) (denatured with 5% isopropanol and 5% methanol)
Catalog # 71002-398
Supplier:  Decon Labs
Supplier Number:  12R1001
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    1 gal.
  • Cat. No.:
    71002-398
  • Supplier no.:
    12R1001

Specifications

Specification Test Results

  • Assay:
    200 proof
  • Ethyl alcohol:
    89-91 (% v/v)
  • Methyl alcohol:
    4.0-5.0 (% v/v)
  • Isopropyl alcohol:
    4.5-5.5 (% v/v)
  • Specific gravity @25C:
    0.782-0.793
  • Water:
    0.01 (% v/v) max.

Specification Test Results

About this item

 :
 :46.07 g/mol
Storage Temperature:Ambient
EINECS:200-578-6
UN:1170
ADR:3,II
 :GHS02
CAS Number:64-17-5
Formula:H₃CCH₂OH
MDL Number:MFCD00003568