Span® 85
: Arlacel 85, Sorbitan trioleate
:26266-58-0
:
:C₆₀H₁₀₈O₈
:MFCD00133820
:∼100 °C (1013 hPa)
:0.95 g/cm³ (20 °C)
:957.51 g/mol
:>110 °C
:Ambient
:247-569-3
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.