Polyethylene glycol 2,000
:25322-68-3
:
:H(OCH₂CH₂)nOH
:MFCD00081839
:2000 g/mol
:49 - 52 °C
:1.21 g/cm³ (20 °C)
:250 °C
:Ambient
:500-038-2
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.